Fumaric acid monoethyl ester, 95%
Fumaric acid monoethyl ester, 95%
Fumaric acid monoethyl ester, 95%
Fumaric acid monoethyl ester, 95%
Acros Organics

Fumaric acid monoethyl ester, 95%

CAS: 2459-05-4 | C6H8O4 | 144.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 156200250
also known as 15620
HSN 29171990
Price (INR)
21,186.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
21,186.00
Each
Chemical Identifiers
CAS2459-05-4
IUPAC Name(2E)-4-ethoxy-4-oxobut-2-enoic acid
Molecular FormulaC6H8O4
InChI KeyXLYMOEINVGRTEX-ONEGZZNKSA-N
SMILESCCOC(=O)\C=C\C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=94 %
Appearance (Color)White to beige
Infrared spectrumConforms
Melting point64°C to 69°C
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only