Nunc™ EasYFlask™ Flasks with Nunclon™ Supra Surface
Thermo Scientific™
Nunc™ EasYFlask™ Flasks with Nunclon™ Supra Surface
Experience optimal performance with Thermo Scientific Nunc EasYFlask flasks with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCulture AreaClosure Type
156380175 cm²Filter
15637225 cm²Filter
15637425 cm²Solid
15637675 cm²Filter
15637875 cm²Solid
156382175 cm²Solid
Catalog number 156380
Price (INR)
12,959.00
Case of 30
Culture Area:
175 cm²
Closure Type:
Filter
Experience optimal performance with Thermo Scientific Nunc EasYFlask flasks with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells. The ergonomic design with angled neck and wide opening allows easy access to the growth surface by pipettes or cell scrapers. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.
Thermo Scientific Nunc vessels with Nunclon Supra surface treatment are designed for culturing fastidious adherent cells. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.
The Nunclon Supra surface supports serum-free and coating-free culture of human mesenchymal stromal cells (MSCs), retaining their morphology and promoting high cell viability. Nunclon Supra cultureware is a great xeno-free solution for cell therapy research applications.
Features of Nunc EasYFlask flasks with Nunclon Supra surface include:
• Consistent lot-to-lot performance
• Sterile with SAL 10-6
• Non-cytotoxic
• Non-pyrogenic
• Biocompatible per USP Class VI for raw materials
• Shelf life: 5 years
For Laboratory Use
Specifications
Closure TypeFilter
DescriptionNunc EasYFlask, 175 cm²
No. per Case30 Flasks
No. per Pack5 Flasks
Bottom ShapeFlat
Culture Area175 cm²
MaterialPolystyrene
Mouth StyleWide
SterilitySterile
Unit SizeCase of 30