4-Nitrobenzaldehyde, 99%
4-Nitrobenzaldehyde, 99%
4-Nitrobenzaldehyde, 99%
Acros Organics

4-Nitrobenzaldehyde, 99%

CAS: 555-16-8 | C7H5NO3 | 151.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Plastic bottle
Catalog number 156991000
HSN 29130090
Price (INR)
13,019.00
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
13,019.00
Each
Chemical Identifiers
CAS555-16-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to brown
Infrared spectrumConforms
Melting point103°C to 107°C
Appearance (Form)Crystalline powder
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only