Gentamicin (10 mg/mL)
Product Image
Gibco™

Gentamicin (10 mg/mL)

Gentamicin sulfate is a water-soluble antibiotic drug originally purified from the fungus Micromonospora purpurea. Gentamicin acts by binding to theRead more
Have Questions?
Catalog NumberQuantity
15710080
also known as 15710-080
20 x 100 mL
Catalog number 15710080
also known as 15710-080
Price (INR)
-
Quantity:
20 x 100 mL
Gentamicin sulfate is a water-soluble antibiotic drug originally purified from the fungus Micromonospora purpurea. Gentamicin acts by binding to the 30S subunit of the bacterial ribosome leading to inhibition protein synthesis and death in susceptible bacteria. Gibco™ Gentamicin is effective against a wide variety of gram-positive and gram-negative bacteria, and is used for the prevention of bacterial contamination on cell cultures. The recommended working concentration ranges from 0.5 to 50 μg/ml. We offer a variety of antibiotics and antimycotics for cell culture applications.

Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.

Dual-Site cGMP Manufacturing
Gibco™ Gentamicin is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ Gentamicin product made in our Grand Island facility (15710-072). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration10 mg/mL, 10 mg/mL
Recommended StorageStorage conditions: 15-30°C
Shipping conditions: Ambient
Shelf life: 24 months from date of manufacture
Sterile FilteredYes
Validated ApplicationPrevention of Cell Culture Contamination
Physical FormLiquid
Quantity20 x 100 mL
TypeGentamicin
Unit SizeEach