Caps for Nunclon™ Delta Flasks
Thermo Scientific™
Caps for Nunclon™ Delta Flasks
Protect your valuable cell culture samples. These caps for Thermo Scientific™ Nunclon™ Delta Flasks are offered in two styles—vent/close, filter—to meet application needs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
15752775 cm² EasY Flask
15889225 cm² EasY Flask
133001175 cm² EasY Flask
15120925 cm² Nunclon Delta Flask
15115225 cm² Nunclon Delta Flask
14445880 cm² Nunclon Delta Flask
147104175/185/500 cm² Nunclon Delta Flask
15824075 cm² EasY Flask
Catalog number 157527
Price (INR)
-
For Use With (Equipment):
75 cm² EasY Flask
Protect your valuable cell culture samples. These caps for Thermo Scientific™ Nunclon™ Delta Flasks are offered in two styles—vent/close (solid) and filter—to meet your application needs.
Vent/close Caps
  • EasY Flask vent/close caps feature a “Y” mark that allows visual verification
  • Vent position easily distinguished even what flasks are stacked in incubators
Filter Caps
  • Filter membrane provides effective barrier against microorganisms
  • Well-defined, hydrophobic filter membrane offers continuous venting with consistent gas exchange and no lot-to-lot variation
  • Chemically robust membrane meets USP class VI standards and is produced without animal-derived substances
Certifications:
  • Sterilized by gamma irradiation and certified to be nonpyrogenic
Ordering Information:
  • Case of 100
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
ClassClass VI
Closure ColorBlue
Closure TypeFilter
DescriptionHDPE closure
For Use With (Equipment)75 cm² EasY Flask
No. per Case100
No. per Pack1
Product TypeFilter cap
Culture Area75 cm2
MaterialHDPE
SterilitySterile
Surface TreatmentNunclon Delta
Unit SizeCase of 100