2-(p-Tolyl)ethylamine, 97%
2-(p-Tolyl)ethylamine, 97%
2-(p-Tolyl)ethylamine, 97%
Acros Organics

2-(p-Tolyl)ethylamine, 97%

CAS: 3261-62-9 | C9H13N | 135.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 157880050
also known as 15788
HSN 29222990
Price (INR)
15,150.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
15,150.00
Each
Chemical Identifiers
CAS3261-62-9
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.5250 to 1.5280 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only