Formanilide, 99+%
Formanilide, 99+%
Formanilide, 99+%
Acros Organics

Formanilide, 99+%

CAS: 103-70-8 | C7H7NO | 121.139 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
500 g
Packaging:
View all
Plastic bottle
Catalog number 158421000
also known as 15842
HSN 29242990
Price (INR)
6,936.00
Each
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
6,936.00
Each
Add to cart
Chemical Identifiers
CAS103-70-8
IUPAC NameN-phenylformamide
Molecular FormulaC7H7NO
InChI KeyDYDNPESBYVVLBO-UHFFFAOYSA-N
SMILESO=CNC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light grey-beige
Appearance (Form)Crystals
Melting point46°C to 49°C
GC>=99.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only