1,12-Dibromododecane, 96%
1,12-Dibromododecane, 96%
1,12-Dibromododecane, 96%
Acros Organics

1,12-Dibromododecane, 96%

CAS: 3344-70-5 | C12H24Br2 | 328.13 g/mol
Have Questions?
Catalog NumberQuantityPackaging
158455000
also known as 15845
500 gGlass bottle
Catalog number 158455000
also known as 15845
HSN 29033920
Price (INR)
115,517.00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
115,517.00
Each
Chemical Identifiers
CAS3344-70-5
IUPAC Name1,12-dibromododecane
Molecular FormulaC12H24Br2
InChI KeyZJJATABWMGVVRZ-UHFFFAOYSA-N
SMILESBrCCCCCCCCCCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=95 %
Appearance (Form)Crystalline solid
Melting point37°C to 42°C
Appearance (Color)Off-white to light brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only