n-Docosane, 99%
n-Docosane, 99%
n-Docosane, 99%
Acros Organics

n-Docosane, 99%

CAS: 629-97-0 | C22H46 | 310.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 158780250
also known as 15878
HSN 29011000
Price (INR)
6,652.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
6,652.00
Each
Chemical Identifiers
CAS629-97-0
IUPAC Namedocosane
Molecular FormulaC22H46
InChI KeyHOWGUJZVBDQJKV-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Infrared spectrumConforms
Melting point43°C to 47°C
Appearance (Color)White
Appearance (Form)Crystalline solid or crystals and/or chunks
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only