Triphenylmethanol, 97%
Triphenylmethanol, 97%
Triphenylmethanol, 97%
Acros Organics

Triphenylmethanol, 97%

CAS: 76-84-6 | C19H16O | 260.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 158910500
HSN 29062910
Price (INR)
4,985.00
Each
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
4,985.00
Each
Chemical Identifiers
CAS76-84-6
IUPAC Nametriphenylmethanol
Molecular FormulaC19H16O
InChI KeyLZTRCELOJRDYMQ-UHFFFAOYSA-N
SMILESOC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Infrared spectrumConforms
GC>=96.0 %
Appearance (Form)Crystalline powder
Melting point160°C to 164°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only