2-Bromo-2-methylpropane, 96%, stabilized
2-Bromo-2-methylpropane, 96%, stabilized
2-Bromo-2-methylpropane, 96%, stabilized
Acros Organics

2-Bromo-2-methylpropane, 96%, stabilized

CAS: 507-19-7 | C4H9Br | 137.02 g/mol
Have Questions?
Catalog NumberQuantityPackaging
159265000500 mLGlass bottle
Catalog number 159265000
HSN 29033919
Price (INR)
28,122.00
Each
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
28,122.00
Each
Chemical Identifiers
CAS507-19-7
IUPAC Name2-bromo-2-methylpropane
Molecular FormulaC4H9Br
InChI KeyRKSOPLXZQNSWAS-UHFFFAOYSA-N
SMILESCC(C)(C)Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow-brown
Refractive index1.4269 to 1.4289 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=95 %
Stabilizerpotassium carbonate
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only