Accessories for Thermo Scientific™ Refrigerated and Heated Bath Circulators
Thermo Scientific™
Accessories for Thermo Scientific™ Refrigerated and Heated Bath Circulators
Enhance the operation of Refrigerated and Heated Bath Circulators with accessories that include bridges, platforms, cooling coils and connectivity solutions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeFor Use With (Equipment)
160-0036Extension BridgeFor A5B, A10B liftplate
160-0007Extension BridgeS15, S21, S30, S45 Liftplate
160-0009TS
also known as 160-0009
Extension BridgeS49 Liftplate
160-0011TS
also known as 160-0011
Lifting PlatformFor S21, S30, S45
160-0012Lifting PlatformFor S15 Heated Bath Circulator
160-0013TS
also known as 160-0013
Lifting PlatformFor S49 Heated Bath Circulator
160-0014Tap Water Cooling CoilSC100, SC150, AC150, AC200 , PC200
160-0015Tap Water Cooling CoilSC100 or SC150 Immersion Circulator
160-0016Tap Water Cooling CoilSC150L with S13, S15, S30, S45, S49
160-0017Tap Water Cooling CoilSC150L Immersion Circulator+clamp
160-0018TS
also known as 160-0018
Adjustable BridgeSC, AC, PC immersion Circulators
160-0027TS
also known as 160-0027
AdapterRS232 Cable for Bath Circulator
160-0028Female M16x1 Adapter (NPTF)Cold/Heated Bath Circulators
160-0029Male M16x1 Adapter (NPTF)Cold/Heated Bath Circulators
160-0045TS
also known as 160-0045
Fluid Displacement BlockFor A10
160-0070Trolley with CastorsFor A40
160-0071Trolley with CastorsFor A28 or A25
160-0075AdapterRS485 Cable for Bath Circulator
160-0077Extension BridgeFor S15, S21, S30, S45
160-0078BridgeSC Controller on Water Baths
160-0088CageSC100 or SC150 immersion circulator
160-0089CageSC150L Immersion Circulator
160-0090Tap Water Cooling CoilSC100/SC150 Circulator with S5P
160-0091Tap Water Cooling CoilSC100/SC150 Circulator with S6T
160-0092Tap Water Cooling CoilSC100/SC150 Circulator with S3
160-0093Tap Water Cooling CoilSC150L Circulator with S7
160-0094Tap Water Cooling CoilAC150 or AC200 controller with S3
160-0105Fluid Displacement BlockFor A25 or A40
160-0106Fluid Displacement BlockFor A28
160-0121Lifting PlatformFor A5B
160-0142Lifting PlatformFor A10B
Catalog number 160-0036
Price (INR)
-
Type:
Extension Bridge
For Use With (Equipment):
For A5B, A10B liftplate
Enhance the operation of Refrigerated and Heated Bath Circulators with Thermo Scientific™ Accessories which include bridges, platforms, cooling coils and connectivity solutions.
  • Warranty varies by country, please contact us for details
Specifications
For Use With (Equipment)For A5B, A10B liftplate
TypeExtension Bridge
Unit SizeEach