Work Area Covers for Thermo Scientific™ Refrigerated and Heated Bath Circulators
Thermo Scientific™
Work Area Covers for Thermo Scientific™ Refrigerated and Heated Bath Circulators
Work Area Covers for Thermo Scientific™ Refrigerated and Heated Bath Circulators include stainless-steel covers, and covers with leveling devices.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
160-0102Water Bath Models S3 or S7
160-0020TS
also known as 160-0020
Water Bath Model S5P
160-0021Water Bath Model S14P
160-0022TS
also known as 160-0022
Water Bath Model H21P
160-0037Water Bath Model S15
160-0038Water Bath Models S21, S30, S45
160-0040Water Bath Model S49
160-0041Water Bath Model A5B
160-0042Water Bath Model A10B
160-0043Water Bath Model A25B
160-0100Water Bath Model A10
160-0101Water Bath Models A28 or A28F
160-0103Water Bath Model S13
Catalog number 160-0102
Price (INR)
-
For Use With (Equipment):
Water Bath Models S3 or S7
Work Area Covers for Thermo Scientific™ Refrigerated and Heated Bath Circulators include stainless-steel covers, and covers with leveling devices.
  • Warranty varies by country, please contact us for details
Specifications
DescriptionWork area cover with leveling device
For Use With (Equipment)Water Bath Models S3 or S7
TypeCover
Unit SizeEach