Search
| Catalog Number | Type | Holds | For Use With (Equipment) |
|---|---|---|---|
| 1600079 | Rack | Depends on insert purchased | A25B, A40, S21, S30 |
| 1600003 | Rack Insert | 100-10 mm Tubes | A5B,A10B A24B,S49,S19T,S14P,S21P |
| 1600004 | Rack Insert | 60-16 mm Tubes | A5B,A10B A24B,S49,S19T,S14P,S21P |
| 1600006 | Rack Insert | Dishes, plates, bags, pouches | A5B,A10B A24B,S49,S19T,S14P,S21P |
| 1600080 | Rack Insert | 55-10 mm Tubes | Cold and Heated Bath Circulators |
| 1600081 | Rack Insert | 32-16 mm Tubes | A25B, A40, S21, S30 |
| 1600082 | Rack Insert | 13-25 mm Tubes | A25B, A40, S21, S30 |
| 1600083 | Rack Insert | Dishes, plates, bags, pouches | A25B, A40, S21, S30 |
| 1600026 | Rack | Depends on insert purchased | S13, S12T, S15 |
| 1600084 | Rack Insert | 39-10 mm Tubes | S13, S12T, S15 |
| 1600085 | Rack Insert | 20-16 mm Tubes | S13, S12T, S15 |
| 1600086 | Rack Insert | 8-25 mm Tubes | S13, S12T, S15 |
| 1600087 | Rack Insert | Dishes, plates, bags, pouches | S13, S12T, S15 |
| 1600066 | Rack Insert | 26-4.2 mm Tubes | GLACIER G50 |
| 1600067 | Rack Insert | 30-2 mm Tubes | GLACIER G50 |
These stainless-steel racks and inserts support a range of tubes and are designed for use with Thermo Scientific™ Refrigerated and Heated Bath Circulators.