Racks and Inserts for Refrigerated and Heated Bath Circulators
Thermo Scientific™
Racks and Inserts for Refrigerated and Heated Bath Circulators
These stainless-steel racks and inserts support a range of tubes and are designed for use with Thermo Scientific™ Refrigerated and Heated Bath Circulators.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeHoldsFor Use With (Equipment)
1600086Rack Insert8-25 mm TubesS13, S12T, S15
1600003Rack Insert100-10 mm TubesA5B,A10B A24B,S49,S19T,S14P,S21P
1600004Rack Insert60-16 mm TubesA5B,A10B A24B,S49,S19T,S14P,S21P
1600006Rack InsertDishes, plates, bags, pouchesA5B,A10B A24B,S49,S19T,S14P,S21P
1600079RackDepends on insert purchasedA25B, A40, S21, S30
1600080Rack Insert55-10 mm TubesCold and Heated Bath Circulators
1600081Rack Insert32-16 mm TubesA25B, A40, S21, S30
1600082Rack Insert13-25 mm TubesA25B, A40, S21, S30
1600083Rack InsertDishes, plates, bags, pouchesA25B, A40, S21, S30
1600026RackDepends on insert purchasedS13, S12T, S15
1600084Rack Insert39-10 mm TubesS13, S12T, S15
1600085Rack Insert20-16 mm TubesS13, S12T, S15
1600087Rack InsertDishes, plates, bags, pouchesS13, S12T, S15
1600066Rack Insert26-4.2 mm TubesGLACIER G50
1600067Rack Insert30-2 mm TubesGLACIER G50
Catalog number 1600086
Price (INR)
-
Type:
Rack Insert
Holds:
8-25 mm Tubes
For Use With (Equipment):
S13, S12T, S15
These stainless-steel racks and inserts support a range of tubes and are designed for use with Thermo Scientific™ Refrigerated and Heated Bath Circulators.
Specifications
For Use With (Equipment)S13, S12T, S15
Holds8-25 mm Tubes
MaterialStainless Steel
TypeRack Insert
Unit SizeEach