2-Amino-4-methoxybenzothiazole, 97%
2-Amino-4-methoxybenzothiazole, 97%
2-Amino-4-methoxybenzothiazole, 97%
Acros Organics

2-Amino-4-methoxybenzothiazole, 97%

CAS: 5464-79-9 | C8H8N2OS | 180.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 160350010
also known as 16035
HSN 29309099
Price (INR)
8,290.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
8,290.00
Each
Chemical Identifiers
CAS5464-79-9
IUPAC Name4-methoxy-1,3-benzothiazol-2-amine
Molecular FormulaC8H8N2OS
InChI KeyYEBCRAVYUWNFQT-UHFFFAOYSA-N
SMILESCOC1=C2N=C(N)SC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point150°C to 153°C
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only