1-Chloro-2,4-dinitrobenzene, 99%
1-Chloro-2,4-dinitrobenzene, 99%
1-Chloro-2,4-dinitrobenzene, 99%
Acros Organics

1-Chloro-2,4-dinitrobenzene, 99%

CAS: 97-00-7 | C6H3ClN2O4 | 202.55 g/mol
Have Questions?
Catalog NumberQuantityPackaging
160511000100 gPlastic bottle
Catalog number 160511000
HSN 29049090
Price (INR)
4,356.00
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
4,356.00
Each
Chemical Identifiers
CAS97-00-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals or mass and/or
Appearance (Form)chunks
Infrared spectrumConforms
GC>=98.5 %
Melting point48°C to 53°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only