n-Hexane, 99+%, for analysis
n-Hexane, 99+%, for analysis
n-Hexane, 99+%, for analysis
Acros Organics

n-Hexane, 99+%, for analysis

CAS: 110-54-3 | C6H14 | 86.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
25 L
Packaging:
View all
Glass Bottle
Catalog number 160780010
HSN 29011000
Price (INR)
11,599.00
Each
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS110-54-3
IUPAC Namehexane
Molecular FormulaC6H14
InChI KeyVLKZOEOYAKHREP-UHFFFAOYSA-N
SMILESCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Iron (Fe)=<0.1 ppm
Lead (Pb)=<0.1 ppm
Magnesium (Mg)=<0.1 ppm
Zinc (Zn)=<0.1 ppm
GC>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only