Product Image
Applied Biosystems™

Bushing, 1/16 plastic

Have Questions?
Catalog NumberQuantity
1611 unit
Catalog number 161
Price (INR)
-
Quantity:
1 unit
This is a spare part for DNA Sequencers.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)310 Genetic Analyzer, 3100 Genetic Analyzer
Product TypeBushing
Quantity1 unit
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store at room temperature.