trans-Stilbene, 96%, Thermo Scientific Chemicals
trans-Stilbene, 96%, Thermo Scientific Chemicals
trans-Stilbene, 96%, Thermo Scientific Chemicals
Acros Organics

trans-Stilbene, 96%, Thermo Scientific Chemicals

trans-Stilbene, 96%, C14H12, CAS Number-103-30-0, bibenzylidine, trans-stilbene, 1,2-diphenylethylene, trans-1,2-diphenylethylene, alpha,beta-diphenylethylene, stilbene, trans-1,2-diphenylethene, bibenzal, e-stilbene, bibenzylidene, 500g, 95% min. (HPLC), 05,II,537
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 161041000
HSN 29029090
Price (INR)
14,061.00
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
14,061.00
Each
Chemical Identifiers
CAS103-30-0
IUPAC Name[(1E)-2-phenylethenyl]benzene
Molecular FormulaC14H12
InChI KeyPJANXHGTPQOBST-VAWYXSNFSA-N
SMILESC(=C/C1=CC=CC=C1)\C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Melting point122°C to 126°C
Appearance (Color)White to light beige
HPLC>=95 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only