1,3,5-Tribromobenzene, 98%
1,3,5-Tribromobenzene, 98%
1,3,5-Tribromobenzene, 98%
Acros Organics

1,3,5-Tribromobenzene, 98%

CAS: 626-39-1 | C6H3Br3 | 314.80 g/mol
Have Questions?
Catalog NumberQuantityPackaging
161081000100 gGlass bottle
Catalog number 161081000
HSN 29039990
Price (INR)
20,286.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
20,286.00
Each
Chemical Identifiers
CAS626-39-1
IUPAC Name1,3,5-tribromobenzene
Molecular FormulaC6H3Br3
InChI KeyYWDUZLFWHVQCHY-UHFFFAOYSA-N
SMILESBrC1=CC(Br)=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to orange-brown
Appearance (Form)Powder
Infrared spectrumConforms
Melting point118°C to 123°C
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only