Product Image
Thermo Scientific™

DRI™ Opiate Drugs of Abuse Calibrators

Achieve high analytical accuracy while eliminating time-consuming steps in reagent preparation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConcentration
1610TS500 ng/mL
1609150 ng/mL
Catalog number 1610TS
Price (INR)
-
Concentration:
500 ng/mL
Achieve high analytical accuracy while eliminating time-consuming steps in reagent preparation when you choose Thermo Scientific™ DRI™ Opiate Drugs of Abuse Calibrators. These ready-to-use assays in liquid form have been field-tested on a wide range of analyzers. They are enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Applications for an array of clinical chemistry analyzers
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Concentration500 ng/mL
DescriptionDRI; Opiate Calibrator 3
Packaging1 Bottle
Quantity1 x 25 mL
TypeCalibrator
Detectable AnalytesOpiate
Sample TypeUrine
Unit SizeEach
Contents & Storage
2°C to 8°C