Quinine monohydrochloride dihydrate, 99%
Quinine monohydrochloride dihydrate, 99%
Quinine monohydrochloride dihydrate, 99%
Acros Organics

Quinine monohydrochloride dihydrate, 99%

CAS: 6119-47-7 | C20H29ClN2O4 | 396.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass bottle
Catalog number 163720250
HSN 29399900
Price (INR)
13,234.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6119-47-7
IUPAC Name(R)-[(2S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol dihydrate hydrochloride
Molecular FormulaC20H29ClN2O4
InChI KeyMPQKYZPYCSTMEI-VQYSEXPNNA-N
SMILESO.O.Cl.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
Sulfated ash=<0.1 %
Appearance (Form)Crystalline powder or needles
Specific optical rotation-245° to -258° (20°C, 589 nm) (c=2, 0.1 M HCl)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only