5-Vinyl-2-norbornene, 98%, stabilized
5-Vinyl-2-norbornene, 98%, stabilized
5-Vinyl-2-norbornene, 98%, stabilized
Acros Organics

5-Vinyl-2-norbornene, 98%, stabilized

CAS: 3048-64-4 | C9H12 | 120.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
25 mL
2.5 L
Packaging:
View all
Glass bottle
Catalog number 164865000
also known as 16486
HSN 29021900
Price (INR)
19,056.00
Each
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
19,056.00
Each
Chemical Identifiers
CAS3048-64-4
IUPAC Name5-ethenylbicyclo[2.2.1]hept-2-ene
Molecular FormulaC9H12
InChI KeyINYHZQLKOKTDAI-UHFFFAOYNA-N
SMILESC=CC1CC2CC1C=C2
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<50 APHA
Infrared spectrumConforms
Refractive index1.4800 to 1.4820 (20°C, 589 nm)
Appearance (Form)Clear liquid
GC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only