Search
Search

| Catalog Number | Length (Metric) | Diameter (Metric) | Particle Size | Product Type |
|---|---|---|---|---|
| 164941 | 250 mm | 0.075 mm | 2 μm | Analytical Column |
| 164568 | 150 mm | 0.075 mm | 3 μm | Analytical Column |
| 164569 | 250 mm | 0.075 mm | 3 μm | Analytical Column |
| 164570 | 500 mm | 0.075 mm | 3 μm | Analytical Column |
| 164572 | 150 mm | 1 mm | 3 μm | Analytical Column |
| 160434 | 5 mm | 1 mm | 5 μm | Trap Cartridge |
| 164213 | 150 mm | 0.2 mm | 5 μm | Trap Column |
| 164199 | 150 mm | 0.1 mm | 5 μm | Trap Column |
| 164537 | 150 mm | 0.3 mm | 2 μm | Analytical Column |
| 164534 | 150 mm | 0.075 mm | 2 μm | Analytical Column |
| 164560 | 50 mm | 0.3 mm | 2 μm | Analytical Column |
| 164535 | 150 mm | 0.075 mm | 3 μm | Trap Column |
| 164942 | 500 mm | 0.075 mm | 2 μm | Analytical Column |
| 164454 | 50 mm | 1 mm | 2 μm | Analytical Column, NanoViper |
| 164946 | 20 mm | 75 μm | 3 μm | Trap Column |
| 164943 | 150 mm | 0.05 mm | 2 μm | Analytical Column |
| 164940 | 150 mm | 0.075 mm | 2 μm | Analytical Column |
Feature high loading capacity for exceptional suitability in the analysis of low abundant peptides in complex proteomics samples with Thermo Scientific™ PepMap™ 100 C18 LC Columns. PepMap LC columns offer lengths targeting fast or high resolution peptide separation.
Ideal for high-resolution peptide mapping of tryptic, natural, and synthetic peptides as well as LC-MS/MS peptide mapping for protein identification, biomarker discovery, and systems biology.
PepMap nanocolumns are also available pre-assembled with Thermo Scientific™ Dionex™ nanoViper™ Fingertight Fitting for a dead-volume-free connection to 1000 bar.
About Nano, capillary, and micro Acclaim PepMap columns
Nano (75μm), capillary (300μm) and micro (1000μm) columns are ideal for protein and peptide analyses. Sample amount is often limited and smaller I.D. columns can increase in sensitivity in sample-limited situations. While nano LC columns are often used in proteomics, the concentrating effects of capillary and microcolumns are beneficial for metabolomics or for facilitating LC-MS interfacing. Thermo Scientific Acclaim PepMap columns are offered in nano, capillary, and microcolumn in a variety of chemistries for peptides and protein analysis.
Use PepMap Trap columns for desalting and preconcentration of peptides before LC separation with MS detection. The trap columns provide the highest efficiencies for one-dimensional peptide mapping experiments and 2D-LC analyses.