Phenyl salicylate, 99%
Phenyl salicylate, 99%
Phenyl salicylate, 99%
Acros Organics

Phenyl salicylate, 99%

Phenyl salicylate, 99%, C13H10O3, CAS Number-118-55-8, salol, salphenyl, phenyl salicylate, 2-phenoxycarbonylphenol, salicylic acid, phenyl ester, benzoic acid, 2-hydroxy-, phenyl ester, phenyl-2-hydroxybenzoate, musol, phenol salicylate, 2-hydroxybenzoic acid phenyl ester, 250g, 98.5% min. (GC) | CAS: 118-55-8 | C13H10O3 | 214.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 165082500
also known as 16508
HSN 29182390
Price (INR)
5,681.00
Each
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
5,681.00
Each
Chemical Identifiers
CAS118-55-8
IUPAC Namephenyl 2-hydroxybenzoate
Molecular FormulaC13H10O3
InChI KeyZQBAKBUEJOMQEX-UHFFFAOYSA-N
SMILESOC1=CC=CC=C1C(=O)OC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Melting point40°C to 44°C
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only