L(+)-Glutamic acid hydrochloride, 99%
L(+)-Glutamic acid hydrochloride, 99%
L(+)-Glutamic acid hydrochloride, 99%
Acros Organics

L(+)-Glutamic acid hydrochloride, 99%

CAS: 138-15-8 | C5H10ClNO4 | 183.588 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
View all
Glass Bottle
Catalog number 165231000
also known as 16523
HSN 29224290
Price (INR)
3,030.00
Each
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (INR)
3,030.00
Each
Chemical Identifiers
CAS138-15-8
IUPAC Namehydrogen 2-aminopentanedioic acid chloride
Molecular FormulaC5H10ClNO4
InChI KeyRPAJSBKBKSSMLJ-UHFFFAOYNA-N
SMILES[H+].[Cl-].NC(CCC(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<0.5 % (105°C, 5 hrs)
Sulfated ash=<0.1 %
Ammonium (NH4)=<200 ppm
Iron (Fe)=<10 ppm
pH1 to 2.5 (2.5 % at 25°C)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only