Tetra Electrophoresis Companion Core
Tetra Electrophoresis Companion Core
Tetra Electrophoresis Companion Core
Invitrogen™
Tetra Electrophoresis Companion Core
Easily swap and run Invitrogen protein gels in Bio-Rad's Mini-PROTEAN™ Tetra Cell tank with the Tetra Electrophoresis Companion Core (a Tetra Electrophoresis Primary Core is also required). Achieve high-performance separations, fast run times, and run up to four gels using both cores.
Have Questions?
Catalog NumberQuantity
16580381 Ea.
Catalog number 1658038
Price (INR)
15,231.00
Online offer
Ends: 31-Dec-2026
21,758.00
Save 6,527.00 (30%)
Each
Quantity:
1 Ea.
Price (INR)
15,231.00
Online offer
Ends: 31-Dec-2026
21,758.00
Save 6,527.00 (30%)
Each
The Invitrogen Tetra Electrophoresis Companion Core is designed to work seamlessly with our Tetra Electrophoresis Primary Core in Bio-Rad's Mini-PROTEAN Tetra Cell. This companion core enables the running of up to four Invitrogen precast protein gels in the tank at the same time, providing a flexible and efficient solution for routine protein analysis applications such as western blotting and SDS-PAGE. Its intuitive design ensures ease of use, making it accessible for both new and experienced users.
  • Run up to four gels simultaneously—capable of running up to four gels per run to increase sample throughput (with a Primary Core, sold separately)
  • Swap and go—designed to work seamlessly with Bio-Rad's Mini-PROTEAN Tetra Cell, allowing for a smooth transition between gel types
  • Achieve high performance—compatible with Invitrogen precast mini protein gels for high-quality results
  • Access multiple gel chemistries—compatible with all Invitrogen mini gels for diverse applications.
The Tetra Electrophoresis Companion Core enhances the functionality of our Tetra Electrophoresis Primary Core by allowing three or four gels to be run at once, significantly increasing throughput and efficiency in the lab. This combination provides a flexible solution for varying experimental needs, ensuring that your lab can handle high-throughput applications with ease. The Companion Core works seamlessly with our Primary Core, making the transition between different gel types smooth and straightforward.
The Tetra Electrophoresis Companion Core is straightforward to use: simply swap out the Bio-Rad cores from the Bio-Rad Mini-PROTEAN Tetra system for the Invitrogen Tetra primary and companion cores and use Invitrogen mini protein gels following the recommended protocols. The companion core requires the primary core to run. When running only one or two gels, use only the primary core.
With our Tetra core set, running Invitrogen precast gels is easy and fast, giving access to premium protein gels that provide excellent protein separation and resolution. Choose from five unique gel chemistries to tailor your separation to your targeted application, including broad range separation, high molecular weight separations, low molecular weight separation, IEF, and native applications. Invitrogen precast gels are backed by our protein gels performance guarantee, so you can be assured of reliable, reproducible performance, lot after lot.
For Research Use Only.
Specifications
Mode of SeparationMolecular Weight
Quantity1 Ea.
For Use With (Equipment)Bio-Rad Mini-PROTEAN Tetra Cell
TypeElectrophoresis Core
Unit SizeEach