Product Image
Thermo Scientific™

CEDIA™ THC Multi-Level Control Set

Eliminate time-consuming steps yet achieve high analytical accuracy with the ready-to-use Thermo Scientific™ CEDIA™ THC (Cannabinoid) Multi-Level Control Set.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConcentration
166108625 ng/mL
166107850 ng/mL
1661060100 ng/mL
Catalog number 1661086
Price (INR)
-
Concentration:
25 ng/mL
Eliminate time-consuming steps yet achieve high analytical accuracy with the ready-to-use Thermo Scientific™ CEDIA™ THC (Cannabinoid) Drugs of Abuse Control Set in liquid form. These assays have been field-tested on a wide range of analyzers, and they are enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.
  • Largest menu of tests available
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Applications for an array of clinical chemistry analyzers
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Concentration25 ng/mL
DescriptionCEDIA THC 25 Multi-Level Control Set
LevelLow (18.75ng/mL) and High (31.25ng/mL)
Quantity1 x 15 mL Low and 1 x 15 mL High
TypeControl
Detectable Analytes11-nor-9-COOH-D9-THC
Sample TypeUrine
Unit SizeEach