CEDIA™ Cocaine Drugs of Abuse Assays
Actual product may vary
Thermo Scientific™
CEDIA™ Cocaine Drugs of Abuse Assays
Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of CEDIA™ Cocaine Drugs of Abuse Assays.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
16612301 x 495 mL, LC
100095TS1 x 65 mL, MCC
100164133 x 17 mL, Indiko
1000863 x 17 mL, TSC
Catalog number 1661230
Price (INR)
-
Quantity:
1 x 495 mL, LC
Utilize the globally-valued accuracy, precision, lot-to-lot dependability and stable shelf life of CEDIA™ Cocaine Drugs of Abuse Assays, which offer applications for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and CEDIA assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing and developing new assays to respond to ever-emerging drug screening needs.
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Control SetsMGC Multi-Drug Controls
DescriptionCEDIA Cocaine Drugs of Abuse Assays
Quantity1 x 495 mL, LC
Sample TypeUrine
TypeReagent
Detectable AnalytesBenzoylecgonine
DoA CalibratorsMulti-drug Calibrator
Unit SizeEach
Contents & Storage
2°C to 8°C