Product Image
Thermo Scientific™
CEDIA™ THC (Cannabinoid) Drugs of Abuse Assays
Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of CEDIA THC (Cannabinoid) Drugs of Abuse Assays.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
16612561 x 495 mL, LC
1000913 x 17 mL, TSC
100100TS
also known as 100100
1 x 65 mL, MCC
100164333 x 17 mL, Indiko
Catalog number 1661256
Price (INR)
-
Quantity:
1 x 495 mL, LC
Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of Thermo Scientific™ CEDIA™ THC (Cannabinoid) Drugs of Abuse Assays, which offer applications for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and CEDIA assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing and developing new assays to respond to ever-emerging drug screening needs.
  • One of the largest menu of tests available
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Control SetsCEDIA THC Controls
DescriptionCEDIA THC (Cannabinoid) Drugs of Abuse Assays
Quantity1 x 495 mL, LC
Sample TypeUrine
TypeReagent
Detectable Analytes11-nor-Δ9THC-COOH
DoA CalibratorsMulti-level THC Calibrator
Unit SizeEach
Contents & Storage
2°C to 8°C