Product Image
Thermo Scientific™

CEDIA™ Propoxyphene and Methadone Drugs of Abuse Calibrators

Utilize the globally valued accuracy, precision and lot-to-lot dependability of CEDIA™ PPx and Methadone Drugs of Abuse Calibrators.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConcentration
1662856PPX 1200 ng/mL, Methadone 600 ng/mL
1662848PPx 300 ng/mL, Methadone 300 ng/mL
1662864PPX 5000 ng/mL, Methadone 1000 ng/mL
Catalog number 1662856
Price (INR)
-
Concentration:
PPX 1200 ng/mL, Methadone 600 ng/mL

Utilize the globally valued accuracy, precision and lot-to-lot dependability of CEDIA™ PPx and Methadone Drugs of Abuse Calibrators, which are optimized for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and CEDIA assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing on the market.

  • Largest menu of tests available
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Applications for an array of clinical chemistry analyzers
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
ConcentrationPPX 1200 ng/mL, Methadone 600 ng/mL
DescriptionCEDIA Intermediate Calibrator
Packaging1 Bottle
Quantity1 x 5 mL
TypeCalibrator
Detectable AnalytesPPx, Methadone
Sample TypeUrine
Unit SizeEach
Contents & Storage
Do not Freeze; 2° to 8°C