Taurine, 99%
Taurine, 99%
Taurine, 99%
Acros Organics

Taurine, 99%

CAS: 107-35-7 | C2H7NO3S | 125.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
25 g
100 g
500 g
Packaging:
Plastic bottle
Catalog number 166540025
also known as 16654
HSN 29225090
Price (INR)
101,787.00
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
101,787.00
Each
Chemical Identifiers
CAS107-35-7
IUPAC Name2-aminoethane-1-sulfonic acid
Molecular FormulaC2H7NO3S
InChI KeyXOAAWQZATWQOTB-UHFFFAOYSA-N
SMILESNCCS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Sulfated ash=<0.1 %
Appearance (Color)White
Arsenic (As)=<2 ppm
Titration with NaOH>=98.5 %
Chloride (Cl)=<110 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only