DL-Asparagine monohydrate, 98%
DL-Asparagine monohydrate, 98%
DL-Asparagine monohydrate, 98%
Acros Organics

DL-Asparagine monohydrate, 98%

CAS: 3130-87-8 | C4H8N2O3 | 132.12 g/mol
Have Questions?
Catalog NumberQuantity
167120025
also known as 16712
2.5 kg
Catalog number 167120025
also known as 16712
HSN 29241900
Price (INR)
155,285.00
Each
Quantity:
2.5 kg
Request bulk or custom format
Price (INR)
155,285.00
Each
Chemical Identifiers
CAS69833-18-7
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with HClO4>=97.5 %
Appearance (Color)White
Appearance (Form)Crystalline powder
Water10.5 to 13.5 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only