N-Carbethoxy-4-piperidone, 98.5+%
N-Carbethoxy-4-piperidone, 98.5+%
N-Carbethoxy-4-piperidone, 98.5+%
N-Carbethoxy-4-piperidone, 98.5+%
Acros Organics

N-Carbethoxy-4-piperidone, 98.5+%

CAS: 29976-53-2 | C8H13NO3 | 171.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
250 g
Packaging:
Glass bottle
Catalog number 167240500
also known as 16724
HSN 29333990
Price (INR)
9,682.00
Each
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
9,682.00
Each
Add to cart
Chemical Identifiers
CAS29976-53-2
IUPAC Nameethyl 4-oxopiperidine-1-carboxylate
Molecular FormulaC8H13NO3
InChI KeyLUBGFMZTGFXIIN-UHFFFAOYSA-N
SMILESCCOC(=O)N1CCC(=O)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
AppearanceMay darken on storage
Water=<0.5 % (K.F.)
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only