Carbon disulfide, 99.9%, for spectroscopy
Carbon disulfide, 99.9%, for spectroscopy
Carbon disulfide, 99.9%, for spectroscopy
Acros Organics

Carbon disulfide, 99.9%, for spectroscopy

CAS: 75-15-0 | CS2 | 76.131 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
25 mL
2.5 L
Packaging:
Glass Bottle
Catalog number 167715000
also known as 16771
HSN 28131000
Price (INR)
10,344.00
Each
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (INR)
10,344.00
Each
Chemical Identifiers
CAS75-15-0
IUPAC Namemethanedithione
Molecular FormulaCS2
InChI KeyQGJOPFRUJISHPQ-UHFFFAOYSA-N
SMILESS=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
UVat 420 nm A: =<0.01
Residue after evaporation=<0.0005 %
UVat 400 nm A: =<0.1
UVat 390 nm A: =<0.5
UVat 410 nm A: =<0.05
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
RUO – Research Use Only