2-Methylbutane, 99+%, for spectroscopy
2-Methylbutane, 99+%, for spectroscopy
2-Methylbutane, 99+%, for spectroscopy
Acros Organics

2-Methylbutane, 99+%, for spectroscopy

CAS: 78-78-4 | C5H12 | 72.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
Packaging:
Glass bottle
Catalog number 167840010
also known as 16784
HSN 29011000
Price (INR)
15,931.00
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
15,931.00
Each
Chemical Identifiers
CAS78-78-4
IUPAC Name2-methylbutane
Molecular FormulaC5H12
InChI KeyQWTDNUCVQCZILF-UHFFFAOYSA-N
SMILESCCC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
UV1 cm cell vs water
UVat 220 nm A: =<0.07
UVat 240 nm A: =<0.01
Acidity=<0.0005 meq/g
Color scale=<10 APHA
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
RUO – Research Use Only