Tetrachloroethylene, 99+%, spectrophotometric grade
Tetrachloroethylene, 99+%, spectrophotometric grade
Tetrachloroethylene, 99+%, spectrophotometric grade
Acros Organics

Tetrachloroethylene, 99+%, spectrophotometric grade

CAS: 127-18-4 | C2Cl4 | 165.82 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 L
1 L
2.5 L
Packaging:
View all
Metal Drum
Catalog number 167890100
also known as 16789
HSN 29032300
Price (INR)
93,195.00
Each
Quantity:
10 L
Packaging:
Metal Drum
Request bulk or custom format
Price (INR)
93,195.00
Each
Chemical Identifiers
CAS127-18-4
IUPAC Nametetrachloroethene
Molecular FormulaC2Cl4
InChI KeyCYTYCFOTNPOANT-UHFFFAOYSA-N
SMILESClC(Cl)=C(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
GC>=99.0 %
UVat 295 nm A: =<0.3
Residue after evaporation=<0.0005 %
Water=<0.05 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only