Potassium tert-butoxide, 98+%, pure
Potassium tert-butoxide, 98+%, pure
Potassium tert-butoxide, 98+%, pure
Acros Organics

Potassium tert-butoxide, 98+%, pure

CAS: 865-47-4 | C4H9KO | 112.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
2.5 kg
Packaging:
View all
Glass Bottle
Catalog number 168880050
HSN 29051900
Price (INR)
3,693.00
Each
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (INR)
3,693.00
Each
Chemical Identifiers
CAS865-47-4
IUPAC Namepotassium 2-methylpropan-2-olate
Molecular FormulaC4H9KO
InChI KeyLPNYRYFBWFDTMA-UHFFFAOYSA-N
SMILES[K+].CC(C)(C)[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
KOH + K2CO3 (as KOH)=<0.50 %
Appearance (Form)Powder
Appearance (Color)White to off-white
Titration with HCl>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only