Dibenzo-18-crown-6, 98+%
Dibenzo-18-crown-6, 98+%
Dibenzo-18-crown-6, 98+%
Acros Organics

Dibenzo-18-crown-6, 98+%

CAS: 14187-32-7 | C20H24O6 | 360.4 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 169800100
also known as 16980
HSN 29329900
Price (INR)
5,657.00
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
5,657.00
Each
Chemical Identifiers
CAS14187-32-7
IUPAC Name2,5,8,15,18,21-hexaoxatricyclo[20.4.0.0⁹,¹⁴]hexacosa-1(26),9,11,13,22,24-hexaene
Molecular FormulaC20H24O6
InChI KeyYSSSPARMOAYJTE-UHFFFAOYSA-N
SMILESC1COC2=CC=CC=C2OCCOCCOC2=CC=CC=C2OCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Infrared spectrumConforms
Melting point160°C to 164°C
HPLC>=98.0 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only