This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 17005034
also known as 17005-034
Price (INR)
10,492.00
線上優惠
Ends: 31-Mar-2026
17,486.00
Save 6,994.00 (40%)
Each
Quantity:
500 mL
Price (INR)
10,492.00
線上優惠
Ends: 31-Mar-2026
17,486.00
Save 6,994.00 (40%)
Each
with L-Glutamine
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeKeratinocyte Cells, Epithelial Cells
Product TypeSerum Free Medium (SFM)
Quantity500 mL
Shelf Life12 Months
Shipping ConditionRoom Temperature
ClassificationSerum-free
Culture TypeMammalian Cell Culture
FormLiquid
Serum LevelSerum-free
SterilitySterile
Sterilization MethodSterile-filtered
With AdditivesGlutamine, Phenol Red
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C) and protect from light.
Frequently asked questions (FAQs)
For how long can I store Keratinocyte-SFM at 4 degrees C?
The base Keratinocyte-SFM has a shelf life of 12 months when stored at 2 to 8 degrees C, protected from light. Keratinocyte-SFM Supplement has a shelf life of 18 months when stored at -20 to 5 degrees C. The complete media should not be stored for longer than 4 weeks at 4 degrees C