Product Image
Gibco™

Keratinocyte-SFM, with L-glutamine

with L-GlutamineRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
17005059
also known as 17005-059
10 x 500 mL
17005034
also known as 17005-034
500 mL
Catalog number 17005059
also known as 17005-059
Price (INR)
56,436.00
Online offer
Ends: 31-Mar-2026
94,059.00
Save 37,623.00 (40%)
Each
Quantity:
10 x 500 mL
Price (INR)
56,436.00
Online offer
Ends: 31-Mar-2026
94,059.00
Save 37,623.00 (40%)
Each
with L-Glutamine
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeKeratinocyte Cells, Epithelial Cells
Concentration1 X
Product TypeSerum Free Medium (SFM)
Quantity10 x 500 mL
Shelf Life12 Months
ClassificationSerum-free
Culture TypeMammalian Cell Culture
FormLiquid
Serum LevelSerum-free
SterilitySterile
Sterilization MethodSterile-filtered
With AdditivesGlutamine, Phenol Red
Unit SizeEach

Frequently asked questions (FAQs)

For how long can I store Keratinocyte-SFM at 4 degrees C?

The base Keratinocyte-SFM has a shelf life of 12 months when stored at 2 to 8 degrees C, protected from light. Keratinocyte-SFM Supplement has a shelf life of 18 months when stored at -20 to 5 degrees C. The complete media should not be stored for longer than 4 weeks at 4 degrees C

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.