Triethanolamine hydrochloride, +99%, Thermo Scientific Chemicals
Triethanolamine hydrochloride, +99%, Thermo Scientific Chemicals
Triethanolamine hydrochloride, +99%, Thermo Scientific Chemicals
Acros Organics

Triethanolamine hydrochloride, +99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 170055000
also known as 17005
HSN 98020000
Price (INR)
10,652.00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
10,652.00
Each
Chemical Identifiers
CAS637-39-8
IUPAC Namehydrogen 2-[bis(2-hydroxyethyl)amino]ethan-1-ol chloride
Molecular FormulaC6H16ClNO3
InChI KeyHHLJUSLZGFYWKW-UHFFFAOYSA-N
SMILES[H+].[Cl-].OCCN(CCO)CCO
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric>=99.0 %
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Loss on drying=<1.0 % (105°C)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only