N-Carbobenzyloxy-L-aspartic acid, 98%
N-Carbobenzyloxy-L-aspartic acid, 98%
N-Carbobenzyloxy-L-aspartic acid, 98%
N-Carbobenzyloxy-L-aspartic acid, 98%
Acros Organics

N-Carbobenzyloxy-L-aspartic acid, 98%

CAS: 1152-61-0 | C12H13NO6 | 267.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 172050250
also known as 17205
HSN 29224990
Price (INR)
6,060.00
Each
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS1152-61-0
IUPAC Name2-{[(benzyloxy)carbonyl]amino}butanedioic acid
Molecular FormulaC12H13NO6
InChI KeyXYXYXSKSTZAEJW-UHFFFAOYNA-N
SMILESOC(=O)CC(NC(=O)OCC1=CC=CC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder
Infrared spectrumConforms
Titration with NaOH97.5 to 102.5 %
Specific optical rotation+8° to +10° (20°C, 589 nm) (c=1, acetic acid)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only