Product Image
Thermo Scientific™

CEDIA™ LSD Drugs of Abuse Assays

Accurately detect LSD in patient samples with Thermo Scientific™ CEDIA™ LSD Drugs of Abuse Assays, for use on a wide array of clinical chemistry analyzers.
Have Questions?
Catalog NumberQuantity
17321371 x 15 mL
Catalog number 1732137
Price (INR)
-
Quantity:
1 x 15 mL
Accurately detect LSD in patient samples with the Thermo Scientific™ CEDIA™ LSD Drugs of Abuse Assay, for use on a wide array of clinical chemistry analyzers. The scope of drugs of abuse testing is expanding rapidly. Thermo Scientific CEDIA assays continue to meet the challenge. CEDIA immunoassays are valued worldwide for their accuracy, precision, lot-to-lot dependability and stable shelf life. We offer the most extensive menu for drugs of abuse testing and we are continuing to develop new assays to respond to ever-emerging drug screening needs.
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Control SetsMGC Multi-Drug Controls
DescriptionCEDIA LSD Drugs of Abuse Assay
Quantity1 x 15 mL
Sample TypeUrine
TypeReagent
Detectable AnalytesLSD
DoA CalibratorsMulti-drug Calibrator
Unit SizeEach
Contents & Storage
2°C to 8°C