Product Image
Thermo Scientific™

CEDIA™ LSD Drugs of Abuse Calibrators

Utilize the globally valued accuracy, precision and lot-to-lot dependability of CEDIA™ LSD Drugs of Abuse Calibrators.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConcentration
17321530.5 ng/mL
17321611.5 ng/mL
17321963 ng/mL
Catalog number 1732153
Price (INR)
-
Concentration:
0.5 ng/mL
Utilize the globally valued accuracy, precision and lot-to-lot dependability of CEDIA™ LSD Drugs of Abuse Calibrators, which are optimized for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and CEDIA assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing on the market.
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Applications for an array of clinical chemistry analyzers
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Concentration0.5 ng/mL
DescriptionCEDIA™ LSD Cutoff Calibrator
Packaging1 Bottle
Quantity1 x 5 mL
TypeCalibrator
Detectable AnalytesLSD
Sample TypeUrine
Unit SizeEach
Contents & Storage
2°C to 8°C