CEDIA™ MultiDrug Drugs of Abuse Calibrators
Actual product may vary
CEDIA™ MultiDrug Drugs of Abuse Calibrators
CEDIA™ MultiDrug Drugs of Abuse Calibrators
Thermo Scientific™
CEDIA™ MultiDrug Drugs of Abuse Calibrators
Utilize the globally valued accuracy, precision and lot-to-lot dependability of CEDIA™ Multidrug Drugs of Abuse Calibrators.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityType
17322181 x 15 mLIntermediate Calibrator
17303801 x 5 mLIntermediate Calibrator
17303981 x 5 mLHigh Calibrator
17304011 x 5 mLClinical Calibrator
17304281 x 5 mLSecondary Calibrator
17305171 x 15 mLSecondary Calibrator
17322261 x 15 mLHigh Calibrator
18153261 x 5 mLPrimary Calibrator
18153341 x 15 mLPrimary Calibrator
Catalog number 1732218
Price (INR)
-
Quantity:
1 x 15 mL
Type:
Intermediate Calibrator
Utilize the globally valued accuracy, precision and lot-to-lot dependability of CEDIA™ Multidrug Drugs of Abuse Calibrators, which are optimized for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and CEDIA assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing on the market.
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Applications for an array of clinical chemistry analyzers
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
ConcentrationMultianalyte
Packaging1 Bottle
Quantity1 x 15 mL
TypeIntermediate Calibrator
Detectable AnalytesMultiple
Sample TypeUrine
Unit SizeEach
Contents & Storage
Do not Freeze; 2° to 8°C