o-Anisic acid, 99%
o-Anisic acid, 99%
o-Anisic acid, 99%
Acros Organics

o-Anisic acid, 99%

CAS: 579-75-9 | C8H8O3 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 173751000
HSN 29189100
Price (INR)
5,184.00
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
5,184.00
Each
Chemical Identifiers
CAS579-75-9
IUPAC Name2-methoxybenzoic acid
Molecular FormulaC8H8O3
InChI KeyILUJQPXNXACGAN-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
HPLC>=98.5 %
Appearance (Form)Crystalline powder
Melting point99°C to 103°C
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only