DRI™ Gravity-Detect™ Specimen Validity Test Calibrators
Thermo Scientific™
DRI™ Gravity-Detect™ Specimen Validity Test Calibrators
Combat the use of additives and adulterants to mask controlled substances with Thermo Scientific™ DRI™ Gravity-Detect™ Specimen Validity Test Calibrators.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLevel
17551.025
17541.010
Catalog number 1755
Price (INR)
-
Level:
1.025
Combat the use of additives and adulterants to mask controlled substances in urine samples with Thermo Scientific™ DRI™ Gravity-Detect™ Specimen Validity Test Calibrators. Liquid and ready-to-use, these controls can be used with the Thermo Scientific™ DRI™ Gravity-Detect™ Reagent Kit on a variety of general chemistry analyzers. As new adulterant trends sweep through the testing pool, rely on our fast-track product development team to deliver a timely, economical detection solution.
  • Can be performed on a variety of general chemistry analyzers
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionDRI™ Gravity-Detect™ Specimen Validity Test Calibrators
Level1.025
Quantity1 x 25 mL
TypeCalibrator
Detectable AnalytesSpecific Gravity
Sample TypeUrine
Unit SizeEach