N-Methyl-p-anisidine, 98%
N-Methyl-p-anisidine, 98%
N-Methyl-p-anisidine, 98%
Acros Organics

N-Methyl-p-anisidine, 98%

CAS: 5961-59-1 | C8H11NO | 137.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
Packaging:
Glass bottle
Catalog number 177220100
also known as 17722
HSN 29222990
Price (INR)
46,938.00
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
46,938.00
Each
Chemical Identifiers
CAS5961-59-1
IUPAC Name4-methoxy-N-methylaniline
Molecular FormulaC8H11NO
InChI KeyJFXDIXYFXDOZIT-UHFFFAOYSA-N
SMILESCNC1=CC=C(OC)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point33°C to 36°C
Appearance (Color)White to grey-brown
Appearance (Form)Low melting crystalline mass
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only